siRNA Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 1 | 256 | ||
| GenBank mRNA | AK036566.1 | - | 1 | 312 | ||
| AK164833.1 | - | 1 | 376 | |||
| U43585.1 | - | 1 | 263 | |||
| X81634.1 | - | 1 | 38 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ksr1| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 1 | 256 | ||
| GenBank mRNA | AK036566.1 | - | 1 | 312 | ||
| AK164833.1 | - | 1 | 376 | |||
| U43585.1 | - | 1 | 263 | |||
| X81634.1 | - | 1 | 38 | |||