siRNA Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 16 | 2337 | ||
| GenBank mRNA | AK137532.1 | - | 15 | 2074 | ||
| U43585.1 | - | 16 | 2344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ksr1| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013571.2 | NP_038599.1 | 16 | 2337 | ||
| GenBank mRNA | AK137532.1 | - | 15 | 2074 | ||
| U43585.1 | - | 16 | 2344 | |||