siRNA Details
| Gene Symbol: GDF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004962.3 | NP_004953.1 | 3 | 2311 | ||
| GenBank mRNA | BC028237.1 | - | 3 | 1741 | 5245100 | |
| D49492.1 | - | 3 | 2149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GDF10| Gene Symbol: GDF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004962.3 | NP_004953.1 | 3 | 2311 | ||
| GenBank mRNA | BC028237.1 | - | 3 | 1741 | 5245100 | |
| D49492.1 | - | 3 | 2149 | |||