siRNA Details
| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113386.1 | NP_001106857.1 | 7 | 1064 | ||
| NM_013584.2 | NP_038612.1 | 6 | 713 | |||
| GenBank mRNA | AK145864.1 | - | 7 | 1065 | ||
| AK155524.1 | - | 6 | 940 | |||
| BC031929.1 | - | 6 | 706 | 4159053 | ||
| D17444.1 | - | 6 | 720 | |||
| D26177.1 | - | 6 | 720 | |||
| S73495.1 | - | 6 | 720 | |||
| S73496.1 | - | 6 | 720 | |||

Lifr
Genomic Map