siRNA Details
| Gene Symbol: Slc11a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013612.2 | NP_038640.2 | 7, 8 | 737 | ||
| GenBank mRNA | AK090303.1 | - | 7, 8 | 738 | ||
| AK144237.1 | - | 7, 8 | 720 | |||
| AK171343.1 | - | 6, 7 | 973 | |||
| AK171393.1 | - | 7, 8 | 718 | |||
| BC109137.1 | - | 7, 8 | 651 | 40039390 | ||
| BC109138.1 | - | 7, 8 | 651 | 40039397 | ||
| L13732.1 | - | 7, 8 | 715 | |||
| X75355.1 | - | 7, 8 | 696 | |||

Slc11a1
Genomic Map