siRNA Details
| Gene Symbol: Odf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113213.1 | NP_001106684.1 | 8, 9 | 1175 | ||
| NM_001113214.1 | NP_001106685.1 | 8, 9 | 1175 | |||
| NM_001177661.1 | NP_001171132.1 | 7, 8 | 1021 | |||
| NM_013615.3 | NP_038643.1 | 8, 9 | 1148 | |||
| GenBank mRNA | AF000968.1 | - | 6, 7 | 995 | ||
| AF034105.1 | - | 8, 9 | 1111 | |||
| AK148523.1 | - | 8, 9 | 1088 | |||
| AK165588.1 | - | 8, 9 | 1079 | |||
| AK168415.1 | - | 8, 9 | 1077 | |||
| BC057001.1 | - | 9, 10 | 1232 | 6827260 | ||

Odf2
Genomic Map