siRNA Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113374.1 | NP_001106845.1 | 4 | 422 | ||
| NM_013826.3 | NP_038854.2 | 5 | 544 | |||
| GenBank mRNA | AF091872.1 | - | 5 | 495 | ||
| AK002719.1 | - | 4 | 390 | |||
| AK010112.1 | - | 5 | 524 | |||
| AK078782.1 | - | 4 | 390 | |||
| BC004631.1 | - | 4 | 393 | |||
| BC024371.1 | - | 5 | 504 | 4983231 | ||

Mocs2
Genomic Map