siRNA Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB221628.1 | - | 1 | 852 | ||
AF191211.1 | - | 1 | 852 | |||
AF191212.1 | - | 1 | 852 | |||
BC068150.1 | - | 3 | 1439 | |||
BC128307.1 | - | 1 | 847 | |||
BC128308.1 | - | 1 | 848 |

More Information
Other Species Targeted

Panther Classification:
