siRNA Details
| Gene Symbol: Pde1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009978.1 | NP_001009978.1 | 7, 8 | 798 | ||
| NM_001009979.1 | NP_001009979.1 | 7, 8 | 798 | |||
| NM_001159582.1 | NP_001153054.1 | 9, 10 | 1300 | |||
| NM_016744.4 | NP_058024.2 | 8, 9 | 1095 | |||
| GenBank mRNA | AF159298.1 | - | 7, 8 | 798 | ||
| AK014810.1 | - | 7, 8 | 795 | |||
| AK043647.1 | - | 8, 9 | 1096 | |||
| AK050004.1 | - | 9, 10 | 1162 | |||
| AY845863.1 | - | 8, 9 | 847 | |||
| AY845864.1 | - | 7, 8 | 798 | |||
| BC090628.1 | - | 8, 9 | 1063 | 6842675 | ||
| U56649.1 | - | 9, 10 | 1031 | |||

Pde1a
Genomic Map