siRNA Details
| Gene Symbol: Pde1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009978.1 | NP_001009978.1 | 12 | 1266 | ||
| NM_001009979.1 | NP_001009979.1 | 12 | 1266 | |||
| NM_001159582.1 | NP_001153054.1 | 14 | 1768 | |||
| NM_016744.4 | NP_058024.2 | 13 | 1563 | |||
| GenBank mRNA | AF159298.1 | - | 12 | 1266 | ||
| AK014810.1 | - | 12 | 1262 | |||
| AK043647.1 | - | 13 | 1564 | |||
| AK050004.1 | - | 14 | 1630 | |||
| AY845863.1 | - | 13 | 1315 | |||
| AY845864.1 | - | 12 | 1266 | |||
| BC090628.1 | - | 13 | 1531 | 6842675 | ||
| U56649.1 | - | 14 | 1499 | |||

Pde1a
Genomic Map