siRNA Details
| Gene Symbol: Itgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016780.2 | NP_058060.2 | 4 | 649 | ||
| GenBank mRNA | AF026509.1 | - | 4 | 593 | ||
| AK157958.1 | - | 4 | 595 | |||
| AY541673.1 | - | 4 | 563 | |||
| BC125518.1 | - | 4 | 649 | 40130033 | ||
| BC125520.1 | - | 4 | 649 | 40130035 | ||

Itgb3
Genomic Map