siRNA Details
| Gene Symbol: Nedd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111324.2 | NP_001104794.1 | 8 | 4375 | ||
| NM_017464.5 | NP_059492.3 | 7 | 4156 | |||
| GenBank mRNA | AK046357.1 | - | 7 | 4122 | ||
| AK083374.1 | - | 8 | 4305 | |||
| AV367231.2 | - | 1 | 400 | |||
| BC004696.1 | - | 3 | 2424 | |||

Nedd9
Genomic Map