siRNA Details
| Gene Symbol: Myo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019472.2 | NP_062345.2 | 3 | 603 | ||
| GenBank mRNA | AJ249706.2 | - | 3 | 227 | ||
| AK028552.1 | - | 2 | 512 | |||
| AK089482.1 | - | 3 | 607 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo10| Gene Symbol: Myo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019472.2 | NP_062345.2 | 3 | 603 | ||
| GenBank mRNA | AJ249706.2 | - | 3 | 227 | ||
| AK028552.1 | - | 2 | 512 | |||
| AK089482.1 | - | 3 | 607 | |||