siRNA Details
| Gene Symbol: Bloc1s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019788.3 | NP_062762.1 | 5 | 1181 | ||
| GenBank mRNA | AI596295.1 | - | 1 | 24 | ||
| AK038381.1 | - | 5 | 1159 | |||
| AK077720.1 | - | 5 | 1163 | |||
| AK147116.1 | - | 5 | 1184 | |||
| AK170964.1 | - | 5 | 1112 | |||
| BC016554.1 | - | 5 | 1160 | |||
| BC085260.1 | - | 5 | 1150 | 6437935 | ||

Bloc1s6
Genomic Map