siRNA Details
| Gene Symbol: SNU13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003796.1 | NP_001003796.1 | 3 | 1262 | ||
| NM_005008.3 | NP_004999.1 | 4 | 1494 | |||
| GenBank mRNA | BC019282.2 | - | 2 | 1150 | ||
| BC095439.1 | - | 3 | 1231 | 30719732 | ||
| BI770951.1 | - | 1 | 203 | |||
| D50420.1 | - | 3 | 1243 | |||

SNU13
Genomic Map