siRNA Details
| Gene Symbol: Kif21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039472.1 | NP_001034561.1 | 2 | 502 | ||
| GenBank mRNA | AF202893.1 | - | 2 | 480 | ||
| AK089400.1 | - | 2 | 450 | |||
| BY742264.1 | - | 2 | 472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif21b| Gene Symbol: Kif21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039472.1 | NP_001034561.1 | 2 | 502 | ||
| GenBank mRNA | AF202893.1 | - | 2 | 480 | ||
| AK089400.1 | - | 2 | 450 | |||
| BY742264.1 | - | 2 | 472 | |||