siRNA Details
| Gene Symbol: Kcnj10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039484.1 | NP_001034573.1 | 2 | 483 | ||
| GenBank mRNA | AB039879.1 | - | 2 | 340 | ||
| AF322631.1 | - | 2 | 292 | |||
| AK044640.1 | - | 2 | 407 | |||
| AK048864.1 | - | 2 | 486 | |||
| AY374423.1 | - | 1 | 250 | |||
| BC099932.1 | - | 2 | 426 | |||
| BC141088.1 | - | 2 | 306 | 9055700 | ||

Kcnj10
Genomic Map