siRNA Details
| Gene Symbol: Kcnj10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039484.1 | NP_001034573.1 | 2 | 5294 | ||
| GenBank mRNA | AF322631.1 | - | 3 | 2323 | ||
| AK031990.1 | - | 1 | 2241 | |||
| BC037038.1 | - | 1 | 1761 | |||
| BC099932.1 | - | 3 | 5158 | |||

Kcnj10
Genomic Map