siRNA Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122733.1 | NP_001116205.1 | 4 | 703 | ||
| NM_021099.3 | NP_066922.2 | 4 | 699 | |||
| GenBank mRNA | AK046795.1 | - | 4 | 695 | ||
| AK047010.1 | - | 4 | 699 | |||
| AY536430.1 | - | 4 | 662 | |||
| AY536431.1 | - | 4 | 662 | |||
| BC026713.1 | - | 4 | 682 | |||
| BC052457.1 | - | 4 | 674 | 6830637 | ||
| BC075716.1 | - | 4 | 688 | 3673641 | ||
| Y00864.1 | - | 4 | 666 | |||

Kit
Genomic Map