siRNA Details
| Gene Symbol: Pfkm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163487.1 | NP_001156959.1 | 16 | 1614 | ||
| NM_001163488.1 | NP_001156960.1 | 16 | 1550 | |||
| NM_021514.4 | NP_067489.3 | 15 | 1567 | |||
| GenBank mRNA | AK002711.2 | - | 16 | 1517 | ||
| AK049773.1 | - | 16 | 1522 | |||
| BC005526.1 | - | 15 | 1531 | 3495173 | ||

Pfkm
Genomic Map