siRNA Details
| Gene Symbol: Kcnk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021542.4 | NP_067517.1 | 5 | 1664 | ||
| GenBank mRNA | AF259395.1 | - | 5 | 1334 | ||
| AF319542.1 | - | 5 | 1403 | |||
| AK085330.1 | - | 5 | 1665 | |||
| AK132172.1 | - | 5 | 1692 | |||
| AK172608.1 | - | 5 | 1665 | |||
| BC034012.1 | - | 5 | 1666 | 4239113 | ||
| BC058164.1 | - | 5 | 1655 | 5323480 | ||

Kcnk5
Genomic Map