siRNA Details
| Gene Symbol: Kng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102411.1 | NP_001095881.1 | 6 | 889 | ||
| NM_001102412.1 | NP_001095882.1 | 6 | 889 | |||
| NM_023125.3 | NP_075614.1 | 6 | 889 | |||
| GenBank mRNA | AK005547.1 | - | 6 | 854 | ||
| AY462058.1 | - | 6 | 769 | |||
| AY660571.1 | - | 6 | 796 | |||
| BC018158.1 | - | 6 | 841 | 4162960 | ||
| BC108936.1 | - | 6 | 845 | 40039851 | ||
| D84415.1 | - | 6 | 769 | |||
| D84435.1 | - | 6 | 769 | |||

Kng1
Genomic Map