siRNA Details
| Gene Symbol: Kng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023125.3 | NP_075614.1 | 10, 11 | 1344 | ||
| GenBank mRNA | AK005547.1 | - | 10, 11 | 1309 | ||
| BC018158.1 | - | 10, 11 | 1296 | 4162960 | ||
| D84415.1 | - | 10, 11 | 1224 | |||

Kng1
Genomic Map