siRNA Details
| Gene Symbol: PIK3CD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005026.3 | NP_005017.3 | 4, 5 | 586 | ||
| XM_006710687.2 | XP_006710750.1 | 4, 5 | 646 | |||
| XM_006710689.2 | XP_006710752.1 | 3, 4 | 500 | |||
| XM_017001476.1 | XP_016856965.1 | 4, 5 | 692 | |||
| XM_017001477.1 | XP_016856966.1 | 5, 6 | 629 | |||
| XM_017001478.1 | XP_016856967.1 | 5, 6 | 789 | |||
| XM_017001479.1 | XP_016856968.1 | 4, 5 | 658 | |||
| XM_017001480.1 | XP_016856969.1 | 3, 4 | 500 | |||
| XM_017001481.1 | XP_016856970.1 | 3, 4 | 573 | |||
| XM_017001482.1 | XP_016856971.1 | 3, 4 | 495 | |||
| XM_017001483.1 | XP_016856972.1 | 3, 4 | 485 | |||
| XM_017001484.1 | XP_016856973.1 | 4, 5 | 692 | |||
| XM_017001485.1 | XP_016856974.1 | 3, 4 | 485 | |||
| GenBank mRNA | AK314191.1 | - | 4, 5 | 587 | ||
| BC132919.1 | - | 3, 4 | 500 | 40146941 | ||
| BC132921.1 | - | 3, 4 | 500 | 40146943 | ||
| BC144264.1 | - | 3, 4 | 500 | 9052784 | ||
| BC150297.1 | - | 4, 5 | 533 | 8860364 | ||
| CN313981.1 | - | 4, 5 | 586 | |||
| DQ263594.1 | - | 2, 3 | 378 | |||
| DQ263595.1 | - | 2, 3 | 378 | |||
| DQ263596.1 | - | 2, 3 | 378 | |||
| JN190435.1 | - | 3, 4 | 450 | |||
| U57843.1 | - | 3, 4 | 493 | |||
| U86453.1 | - | 4, 5 | 573 | |||
| Y10055.2 | - | 4, 5 | 574 | |||

PIK3CD
Genomic Map