siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153103.2 | NP_694743.2 | 4 | 530 | ||
| GenBank mRNA | AB074017.1 | - | 2 | 160 | ||
| AK029250.1 | - | 4 | 457 | |||
| AK173007.1 | - | 3 | 237 | |||
| BC029286.1 | - | 4 | 517 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif1c| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153103.2 | NP_694743.2 | 4 | 530 | ||
| GenBank mRNA | AB074017.1 | - | 2 | 160 | ||
| AK029250.1 | - | 4 | 457 | |||
| AK173007.1 | - | 3 | 237 | |||
| BC029286.1 | - | 4 | 517 | |||