siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB074017.1 | - | 9 | 884 | ||
| AK173007.1 | - | 10 | 961 | |||
| BC029286.1 | - | 11 | 1241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif1c| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB074017.1 | - | 9 | 884 | ||
| AK173007.1 | - | 10 | 961 | |||
| BC029286.1 | - | 11 | 1241 | |||