siRNA Details
| Gene Symbol: Kif1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153103.2 | NP_694743.2 | 20 | 2154 | ||
| GenBank mRNA | AB074017.1 | - | 18 | 1784 | ||
| AK046574.1 | - | 15 | 1395 | |||
| AK173007.1 | - | 19 | 1861 | |||
| BC006188.1 | - | 2 | 102 | |||
| BC029286.1 | - | 20 | 2141 | |||

Kif1c
Genomic Map