siRNA Details
| Gene Symbol: Nrxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198587.3 | NP_001185516.2 | 7, 8 | 2735 | ||
| NM_172544.3 | NP_766132.2 | 6, 7 | 1625 | |||
| GenBank mRNA | AK032126.1 | - | 6, 7 | 1617 | ||
| AK129205.1 | - | 5, 6 | 1498 | |||
| BC060719.1 | - | 8, 9 | 2310 | 6406001 | ||

Nrxn3
Genomic Map