siRNA Details
| Gene Symbol: Nrxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198587.3 | NP_001185516.2 | 10 | 3049 | ||
| NM_172544.3 | NP_766132.2 | 9 | 1939 | |||
| GenBank mRNA | AK032126.1 | - | 9 | 1931 | ||
| AK129205.1 | - | 8 | 1812 | |||
| BC060719.1 | - | 11 | 2624 | 6406001 | ||

Nrxn3
Genomic Map