siRNA Details
Gene Symbol: Edn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007902.2 | NP_031928.2 | 5 | 1202 | ||
GenBank mRNA | BC037042.1 | - | 5 | 1431 | 4512195 | |
X59556.1 | - | 5 | 1191 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Edn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007902.2 | NP_031928.2 | 5 | 1202 | ||
GenBank mRNA | BC037042.1 | - | 5 | 1431 | 4512195 | |
X59556.1 | - | 5 | 1191 |