siRNA Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 7, 8 | 756 | ||
| GenBank mRNA | AF035527.1 | - | 6, 7 | 754 | ||
| AK028651.1 | - | 7, 8 | 727 | |||
| BC005520.1 | - | 8, 9 | 673 | 3494644 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehf| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 7, 8 | 756 | ||
| GenBank mRNA | AF035527.1 | - | 6, 7 | 754 | ||
| AK028651.1 | - | 7, 8 | 727 | |||
| BC005520.1 | - | 8, 9 | 673 | 3494644 | ||