siRNA Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 9 | 1119 | ||
| GenBank mRNA | AF035527.1 | - | 8 | 1117 | ||
| AK028651.1 | - | 9 | 1090 | |||
| BC005520.1 | - | 10 | 1036 | 3494644 | ||
| BC008249.1 | - | 2 | 312 | |||
| CF424908.1 | - | 2 | 251 | |||

Ehf
Genomic Map