siRNA Details
| Gene Symbol: Endog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007931.1 | NP_031957.1 | 2 | 610 | ||
| GenBank mRNA | AB012108.1 | - | 2 | 610 | ||
| AK144459.1 | - | 2 | 642 | |||
| BC030177.1 | - | 2 | 619 | 5029633 | ||
| X99394.1 | - | 2 | 106 | |||

Endog
Genomic Map