siRNA Details
| Gene Symbol: Eps15l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122832.1 | NP_001116304.1 | 7 | 455 | ||
| NM_001289859.1 | NP_001276788.1 | 7 | 455 | |||
| NM_007944.3 | NP_031970.2 | 7 | 455 | |||
| GenBank mRNA | AK036662.1 | - | 7 | 421 | ||
| AK036728.1 | - | 7 | 433 | |||
| AK146781.1 | - | 7 | 433 | |||
| AK152602.1 | - | 7 | 433 | |||
| BB855263.1 | - | 7 | 458 | |||
| BC015259.1 | - | 7 | 436 | 4238768 | ||
| U29156.1 | - | 7 | 442 | |||

Eps15l1
Genomic Map