siRNA Details
| Gene Symbol: SEL1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244984.1 | NP_001231913.1 | 2 | 204 | ||
| NM_005065.5 | NP_005056.3 | 2 | 204 | |||
| XM_005267988.2 | XP_005268045.1 | 2 | 150 | |||
| XM_005267989.3 | XP_005268046.1 | 2 | 207 | |||
| GenBank mRNA | AB020335.1 | - | 2 | 132 | ||
| AF052059.1 | - | 2 | 180 | |||
| AK075511.1 | - | 2 | 183 | |||
| AY358651.1 | - | 2 | 176 | |||
| BP358857.1 | - | 2 | 204 | |||

SEL1L
Genomic Map