siRNA Details
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271587.1 | NP_001258516.1 | 5 | 676 | ||
| NM_001271588.1 | NP_001258517.1 | 5 | 607 | |||
| NM_001271595.1 | NP_001258524.1 | 5 | 681 | |||
| NM_007945.3 | NP_031971.2 | 6 | 682 | |||
| GenBank mRNA | AK147877.1 | - | 6 | 610 | ||
| AK148424.1 | - | 6 | 598 | |||
| AK149683.1 | - | 5 | 470 | |||
| AK155022.1 | - | 5 | 442 | |||
| AK166156.1 | - | 6 | 517 | |||
| BC016890.1 | - | 5 | 693 | 4240899 | ||
| L21671.1 | - | 5 | 597 | |||

Eps8
Genomic Map