siRNA Details
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271587.1 | NP_001258516.1 | 7, 8 | 931 | ||
| NM_001271588.1 | NP_001258517.1 | 7, 8 | 862 | |||
| NM_001271595.1 | NP_001258524.1 | 7, 8 | 936 | |||
| NM_007945.3 | NP_031971.2 | 8, 9 | 937 | |||
| GenBank mRNA | AK147877.1 | - | 8, 9 | 865 | ||
| AK148424.1 | - | 8, 9 | 853 | |||
| AK149683.1 | - | 7, 8 | 725 | |||
| AK155022.1 | - | 7, 8 | 697 | |||
| AK166156.1 | - | 8, 9 | 772 | |||
| BC016890.1 | - | 7, 8 | 948 | 4240899 | ||
| L21671.1 | - | 7, 8 | 852 | |||

Eps8
Genomic Map