siRNA Details
| Gene Symbol: Ptprv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007955.3 | NP_031981.3 | 9 | 1645 | ||
| GenBank mRNA | AF300701.1 | - | 8 | 1122 | ||
| AK156112.1 | - | 9 | 1647 | |||
| U36488.1 | - | 5 | 807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprv| Gene Symbol: Ptprv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007955.3 | NP_031981.3 | 9 | 1645 | ||
| GenBank mRNA | AF300701.1 | - | 8 | 1122 | ||
| AK156112.1 | - | 9 | 1647 | |||
| U36488.1 | - | 5 | 807 | |||