siRNA Details
| Gene Symbol: Esx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007957.2 | NP_031983.2 | 5 | 1216 | ||
| GenBank mRNA | AF004211.1 | - | 4 | 1304 | ||
| AF017734.1 | - | 4 | 933 | |||
| AF017735.1 | - | 5 | 1016 | |||
| AF085715.1 | - | 5 | 1217 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esx1| Gene Symbol: Esx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007957.2 | NP_031983.2 | 5 | 1216 | ||
| GenBank mRNA | AF004211.1 | - | 4 | 1304 | ||
| AF017734.1 | - | 4 | 933 | |||
| AF017735.1 | - | 5 | 1016 | |||
| AF085715.1 | - | 5 | 1217 | |||