siRNA Details
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ001482.1 | - | 5 | 1081 | ||
JQ665270.1 | - | 5 | 1073 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106423, XM_006232249, XM_008760909, XM_008760910, XM_006232250, and others.
Panther Classification:
