siRNA Details
| Gene Symbol: Ptk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130409.1 | NP_001123881.1 | 18, 19 | 1889 | ||
| NM_007982.2 | NP_032008.2 | 17, 18 | 1734 | |||
| GenBank mRNA | AB011499.1 | - | 18, 19 | 1545 | ||
| AK079821.1 | - | 18, 19 | 1986 | |||
| AK220543.1 | - | 17, 18 | 1734 | |||
| BC030180.1 | - | 18, 19 | 1819 | |||
| JN971016.1 | - | 17, 18 | 1461 | |||
| M95408.1 | - | 16, 17 | 1531 | |||

Ptk2
Genomic Map