siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042673.2 | NP_001036138.1 | 8 | 1130 | ||
| NM_001282942.1 | NP_001269871.1 | 7 | 933 | |||
| NM_007985.3 | NP_032011.2 | 8 | 1130 | |||
| GenBank mRNA | AK035488.1 | - | 8 | 884 | ||
| AK042226.1 | - | 8 | 1100 | |||
| AK080697.1 | - | 1111 | ||||
| AK082717.1 | - | 7 | 911 | |||
| AK147025.1 | - | 8 | 1110 | |||
| AK163161.1 | - | 8 | 1131 | |||
| BC038609.1 | - | 7 | 915 | 5375091 | ||
| L08266.1 | - | 8 | 1111 | |||

Fancc
Genomic Map