siRNA Details
| Gene Symbol: Fap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007986.3 | NP_032012.1 | 5, 6 | 538 | ||
| GenBank mRNA | AK051959.1 | - | 5, 6 | 441 | ||
| AK136578.1 | - | 4, 5 | 528 | |||
| BC019190.1 | - | 5, 6 | 442 | 5028579 | ||
| Y10007.1 | - | 5, 6 | 538 | |||

Fap
Genomic Map