siRNA Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007995.3 | NP_032021.1 | 7, 8 | 637 | ||
| GenBank mRNA | AB007813.1 | - | 7, 8 | 529 | ||
| AB222271.1 | - | 6, 7 | 418 | |||
| AK133539.1 | - | 7, 8 | 614 | |||
| AK138752.1 | - | 7, 8 | 584 | |||
| AK159651.1 | - | 7, 8 | 607 | |||
| BC019180.1 | - | 7, 8 | 585 | 5008934 | ||
| CT010383.1 | - | 7, 8 | 499 | |||

Fcna
Genomic Map