siRNA Details
| Gene Symbol: JMJD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114632.1 | NP_001108104.1 | 5 | 585 | ||
| GenBank mRNA | AK000550.1 | - | 5 | 691 | ||
| AK000663.1 | - | 5 | 559 | |||
| AK000814.1 | - | 4 | 492 | |||
| AK225503.1 | - | 5 | 559 | |||
| AK289833.1 | - | 5 | 585 | |||
| AK290051.1 | - | 5 | 554 | |||
| BC025290.1 | - | 5 | 547 | |||
| BC139899.1 | - | 5 | 564 | |||
| Gene Symbol: JMJD7-PLA2G4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198588.1 | NP_001185517.1 | 5 | 585 | ||
| NM_005090.3 | NP_005081.1 | 5 | 585 | |||
| GenBank mRNA | AB209150.1 | - | 5 | 575 | ||
| AF065215.1 | - | 5 | 560 | |||
| AF065216.1 | - | 4 | 636 | |||
| AF121908.1 | - | 5 | 552 | |||
| DQ523799.1 | - | 5 | 553 | |||
| DQ523800.1 | - | 5 | 553 | |||

JMJD7
Genomic Map