siRNA Details
| Gene Symbol: Fgf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008003.2 | NP_032029.1 | 1, 2 | 401 | ||
| GenBank mRNA | AF007268.1 | - | 1, 2 | 401 | ||
| AK017829.1 | - | 1, 2 | 403 | |||
| BC021328.1 | - | 1, 2 | 393 | 5066286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf15| Gene Symbol: Fgf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008003.2 | NP_032029.1 | 1, 2 | 401 | ||
| GenBank mRNA | AF007268.1 | - | 1, 2 | 401 | ||
| AK017829.1 | - | 1, 2 | 403 | |||
| BC021328.1 | - | 1, 2 | 393 | 5066286 | ||