siRNA Details
| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005094.3 | NP_005085.2 | 3 | 476 | ||
| XM_017014222.1 | XP_016869711.1 | 4 | 349 | |||
| GenBank mRNA | AF055899.1 | - | 2 | 191 | ||
| AK027504.1 | - | 2 | 56 | |||
| AK290222.1 | - | 3 | 476 | |||
| AK314681.1 | - | 3 | 316 | |||
| BC065003.1 | - | 3 | 475 | 6023438 | ||

SLC27A4
Genomic Map