siRNA Details
| Gene Symbol: Fpr-rs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008041.2 | NP_032067.2 | 1 | 826 | ||
| GenBank mRNA | BC115534.1 | - | 1 | 822 | ||
| BC115535.1 | - | 1 | 826 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpr-rs4| Gene Symbol: Fpr-rs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008041.2 | NP_032067.2 | 1 | 826 | ||
| GenBank mRNA | BC115534.1 | - | 1 | 822 | ||
| BC115535.1 | - | 1 | 826 | |||
Set Membership: |
|