siRNA Details
| Gene Symbol: Frat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008043.3 | NP_032069.2 | 1 | 860 | ||
| GenBank mRNA | AK015680.1 | - | 1 | 794 | ||
| BC140987.1 | - | 1 | 714 | 9055599 | ||
| U58974.2 | - | 1 | 714 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frat1| Gene Symbol: Frat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008043.3 | NP_032069.2 | 1 | 860 | ||
| GenBank mRNA | AK015680.1 | - | 1 | 794 | ||
| BC140987.1 | - | 1 | 714 | 9055599 | ||
| U58974.2 | - | 1 | 714 | |||