siRNA Details
| Gene Symbol: Fst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301373.1 | NP_001288302.1 | 6 | 1369 | ||
| NM_001301375.1 | NP_001288304.1 | 6 | 1669 | |||
| NM_008046.3 | NP_032072.1 | 6 | 1366 | |||
| GenBank mRNA | AK083556.1 | - | 6 | 1451 | ||
| BC144926.1 | - | 6 | 1193 | 9053474 | ||
| BC145945.1 | - | 6 | 1193 | 40131248 | ||
| X83377.1 | - | 6 | 998 | |||
| Z29532.1 | - | 6 | 1001 | |||
| Gene Symbol: Thrap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC012655.1 | - | 1055 | |||

Fst
Genomic Map